Rhodium(II) acetate dimer (∼46% Rh), vedetön

Toimittaja: Thermo Fisher Scientific
Varoitus

268630010 268632500 268630050
ACRO268630010EA 1400 EUR
ACRO268630010 ACRO268632500 ACRO268630050
Rhodium(II) acetate dimer (∼46% Rh), vedetön
Rhodium(II) acetate dimer
Kaava: C₈H₁₂O₈Rh₂
Sulamispiste: 205 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003538
CAS-numero: 15956-28-2
EINECS: 240-084-8

Order Now

Testitulosten erittely

Appearance Green Crystalline powder
Infrared spectrum Conforms
Assay 46 % (Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR