Palladium 99.9% (trace metals basis), sponge

Toimittaja: Thermo Fisher Scientific
Varoitus

268150010
ACRO268150010EA 402 EUR
ACRO268150010
Palladium 99.9% (trace metals basis), sponge
Palladium
Kaava: Pd
Kiehumispiste: 3167 °C (1013 hPa)
Sulamispiste: 1555 °C
Tiheys: 12,02 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3
EINECS: 231-115-6
UN: 3089
Merck Index: 12,07121

Order Now

Testitulosten erittely

Appearance Grey Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR