L-Dehydroascorbic acid 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

250930010 250930050 250932500
ACRO250930010EA 170 EUR
ACRO250930010 ACRO250930050 ACRO250932500
L-Dehydroascorbic acid 96%
L-Dehydroascorbic acid
Kaava: C₆H₆O₆
Sulamispiste: 228 °C
Storage Temperature: Pakastin
MDL numero: MFCD00066467
CAS-numero: 490-83-5

Order Now

Testitulosten erittely

Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥95.0 % (ELSD)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR