Disodium hydrogen citrate sesquihydrate 99%, pure

Toimittaja: Thermo Fisher Scientific

250240010 250240025 250242500
ACRO250240010EA 78.5 EUR
ACRO250240010 ACRO250240025 ACRO250242500
Disodium hydrogen citrate sesquihydrate 99%, pure
Disodium hydrogen citrate sesquihydrate
Kaava: C₆H₆Na₂O₇·1.5H₂O
Sulamispiste: 149 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150445
CAS-numero: 6132-05-4
EINECS: 242-734-6

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
pH 4.9 to 5.2 (3 % solution)
Titration with HClO4 98.5 to 104.0 %
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR