2-Methyl-5-isopropylaniline (synthetic) ≥94% (by GC)

Toimittaja: Thermo Fisher Scientific
Varoitus

248790010 248790050 248790250
ACRO248790010EA 113 EUR
ACRO248790010 ACRO248790050 ACRO248790250
2-Methyl-5-isopropylaniline (synthetic) ≥94% (by GC)
2-Methyl-5-isopropylaniline
Kaava: C₁₀H₁₅N
Tiheys: 0,95 g/cm³
MDL numero: MFCD00044503
CAS-numero: 2051-53-8

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow to brown liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5450 (20°C, 589 nm)
Water ≤2.5 %
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR