Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)

Toimittaja: Thermo Fisher Scientific

244021000 244022500 244020010
ACRO244021000EA 184 EUR
ACRO244021000 ACRO244022500 ACRO244020010
Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)
Hydroxypropylmethylcellulose
Kaava: C₃H₈O₂·xCH₄O
Tiheys: 1,39 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00131360
CAS-numero: 9004-65-3

Order Now

Testitulosten erittely

Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR