Trichloromethyl chloroformate 99%

Toimittaja: Thermo Fisher Scientific
Vaara

243310100 243311000
ACRO243310100EA 51.9 EUR
ACRO243310100 ACRO243311000
Trichloromethyl chloroformate 99%
Trichloromethyl chloroformate
Kaava: C₂Cl₄O₂
Kiehumispiste: 128 °C (760 mmHg)
Sulamispiste: –57 °C
Tiheys: 1,64 g/cm³
Storage Temperature: Jääkaappi
MDL numero: MFCD00015553
CAS-numero: 503-38-8
EINECS: 207-965-9
UN: 3390
ADR: 6.1,
Merck Index: 13,03368

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR