L(-)-Glukoosi 98%

Toimittaja: Thermo Fisher Scientific

241920010 241922500 241920050 241920250
ACRO241920010EA 74.3 EUR
ACRO241920010 ACRO241922500 ACRO241920050 ACRO241920250
L(-)-Glukoosi 98%
L(-)-Glukoosi
Kaava: C₆H₁₂O₆
Kiehumispiste: 376 °C (1013 hPa)
Sulamispiste: 153…156 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00148913
CAS-numero: 921-60-8
EINECS: 213-068-3

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (10 % in water) Clear colorless
HPLC ≥97.5 % (ELSD)
Specific optical rotation -50° to -54° (20°C, 589 nm) (c=10, H2O, NH3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR