Silikageeli, Pore size: 90 Å 0.035-0.070 mm kromatografiaan

Toimittaja: Thermo Fisher Scientific

241650010 241652500 241650000
ACRO241650010EA 109 EUR
ACRO241650010 ACRO241652500 241650000.
Silikageeli, Pore size: 90 Å 0.035-0.070 mm kromatografiaan
Silikageeli
Kaava: SiO₂
Kiehumispiste: 2230 °C (1013 hPa)
Sulamispiste: 1710 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 7631-86-9
REACH: 01-2119379499-16

Order Now

Testitulosten erittely

Appearance White Powder
Particle size ≤10 % oversized particles
Bulk density 385 to 440 g/l
pH 6.5 to 7.5 (5 % w/w suspension)
Residual water 2 to 4 % (160°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR