Kalium-D-glukonaatti 99%

Toimittaja: Thermo Fisher Scientific

229320010 229322500
ACRO229320010EA 115 EUR
ACRO229320010 ACRO229322500
Kalium-D-glukonaatti 99%
Kalium-D-glukonaatti
Kaava: HOH₂C(CH(OH))₄COOK
Sulamispiste: 178…181 °C
Tiheys: 1,73 g/cm³ (20 °C)
MDL numero: MFCD00064211
CAS-numero: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Assay ≥98.5 % (C6H11KO7) On dried substance
Loss on drying ≤3 % (105°C, 4 h) (Vacuum)
Heavy metals (as Pb) ≤20 ppm
Reducing substances ≤1 % (as Dextrose)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR