Puromycin dihydrochloride hydrate 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

227420100 227420500 227422500
ACRO227420100EA 83.6 EUR
ACRO227420100 ACRO227420500 ACRO227422500
Puromycin dihydrochloride hydrate 99%
Puromycin dihydrochloride hydrate
Kaava: C₂₂H₂₉N₇O₅·2HCl·nH₂O
Sulamispiste: 168…178 °C
Storage Temperature: Pakastin
MDL numero: MFCD00150080
CAS-numero: 58-58-2

Order Now

Testitulosten erittely

Appearance White to light beige Powder
Infrared spectrum Conforms
Water ≤12 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR