Erythromycin 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: (-)-Erythromycin

227330050 227330250 227331000
ACRO227330050EA 79.3 EUR
ACRO227330050 ACRO227330250 ACRO227331000
Erythromycin 98%
Erythromycin
Kaava: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
Sulamispiste: 138…140 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00084654
CAS-numero: 114-07-8
EINECS: 204-040-1

Order Now

Testitulosten erittely

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤6.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥97.5 % (sum A,B and C)
Specific optical rotation -71° to -78° (20°C, 589 nm) (c=2, C2H5OH, 30 minutes) on dry substance


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR