L(-)-Ksyloosi 99+%

Toimittaja: Thermo Fisher Scientific

225990050 225990250 225991000
ACRO225990050EA 58.1 EUR
ACRO225990050 ACRO225990250 ACRO225991000
L(-)-Ksyloosi 99+%
L(-)-Ksyloosi
Kaava: C₅H₁₀O₅
Kiehumispiste: 331 °C (1013 hPa)
Sulamispiste: 150…152 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151096
CAS-numero: 609-06-3
EINECS: 210-174-1

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -18° to -20° (20°C, 589 nm) (c=10, H2O) (after 24 h)
Loss on ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR