(3,4-Dihydroksifenyyli)etikkahappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

225600010 225600050 225600250
ACRO225600010EA 47.1 EUR
ACRO225600010 ACRO225600050 ACRO225600250
(3,4-Dihydroksifenyyli)etikkahappo 98%
(3,4-Dihydroksifenyyli)etikkahappo
Kaava: (C₆H₃(OH)₂)CH₂COOH
Kiehumispiste: 337 °C (1013 hPa)
Sulamispiste: 126…130 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004338
CAS-numero: 102-32-9
EINECS: 203-024-1

Order Now

Testitulosten erittely

Appearance Beige to light brown to purple-grey Powder
Infrared spectrum Conforms
Melting point 127 to 132 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR