Chlorocarbonylsulphenyl chloride 95%

Toimittaja: Thermo Fisher Scientific
Vaara

224340050 224340250
ACRO224340050EA 77.7 EUR
ACRO224340050 ACRO224340250
Chlorocarbonylsulphenyl chloride 95%
Chlorocarbonylsulphenyl chloride
Kaava: CCl₂OS
Kiehumispiste: 98…100 °C (760 mmHg)
Tiheys: 1,58 g/cm³
Leimahduspiste: 62 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00000703
CAS-numero: 2757-23-5
UN: 3265
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR