(S)-(+)-2-Methylbutyric acid 98%

Toimittaja: Thermo Fisher Scientific
Vaara

224190010 224190050 224190250
ACRO224190010EA 109 EUR
ACRO224190010 ACRO224190050 ACRO224190250
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid
Kaava: C₅H₁₀O₂
Kiehumispiste: 78 °C (15 mmHg)
Tiheys: 0,938 g/cm³
Leimahduspiste: 73 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00063166
CAS-numero: 1730-91-2
UN: 3265
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR