Kalium 98%, chunks in mineral oil

Toimittaja: Thermo Fisher Scientific
Vaara

223860100 223861000 223860500 223865000
ACRO223860100EA 54.5 EUR
ACRO223860100 ACRO223861000 ACRO223860500 ACRO223865000
Kalium 98%, chunks in mineral oil
Kalium
Kaava: K
Kiehumispiste: 770 °C (760 mmHg)
Sulamispiste: 63 °C
Tiheys: 0,86 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00133776
CAS-numero: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3,I
Merck Index: 12,07763

Order Now

Testitulosten erittely

Appearance Grey Chunks
Total trace metal impurities ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR