Tina(IV)kloridi-5 H2O 98+%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Tin tetrachloride pentahydrate, Stannic chloride pentahydrate

223691000 223695000
ACRO223691000EA 47.1 EUR
ACRO223691000 ACRO223695000
Tina(IV)kloridi-5 H2O 98+%, extra pure
Tina(IV)kloridi-5 H2O
Kaava: Cl₄Sn·5H₂O
Sulamispiste: 56 °C
Tiheys: 2,2 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149864
CAS-numero: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White to off-white Powder or crystals and/or lumps
Assay ≥98.0 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR