1-Oktyyni 99%

Toimittaja: Thermo Fisher Scientific
Vaara

223510250 223511000
ACRO223510250EA 57.9 EUR
ACRO223510250 ACRO223511000
1-Oktyyni 99%
1-Oktyyni
Kaava: C₈H₁₄
MW: 110.2 g/mol
Kiehumispiste: 127…128 °C (760 mmHg)
Sulamispiste: –80 °C
Tiheys: 0.74 g/cm³
Leimahduspiste: 17 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009546
CAS-numero: 629-05-0
EINECS: 211-069-3
UN: 3295
ADR: 3 II

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR