5-Kloori-1-pentyyni 98%

Toimittaja: Thermo Fisher Scientific
Vaara

223420050 223420250
ACRO223420050EA 73.9 EUR
ACRO223420050 ACRO223420250
5-Kloori-1-pentyyni 98%
5-Kloori-1-pentyyni
Kaava: C₅H₇Cl
Kiehumispiste: 67…69 °C (145 mmHg)
Tiheys: 0,96 g/cm³
Leimahduspiste: 15 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00001014
CAS-numero: 14267-92-6
EINECS: 238-159-5
UN: 1993
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR