Aniliini 99.5%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Aminobenzene, Phenylamine

221730010 221732500 221730025 221730050
ACRO221730010EA 78.2 EUR
ACRO221730010 ACRO221732500 ACRO221730025 ACRO221730050
Aniliini 99.5%, extra pure
Aniliini
Kaava: C₆H₅NH₂
Kiehumispiste: 181…185 °C (760 mmHg)
Sulamispiste: –6,2 °C
Tiheys: 1,0213 g/cm³
Leimahduspiste: 76 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007629
CAS-numero: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5875 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR