Lyijy(II,IV)oksidi 98%

Toimittaja: Thermo Fisher Scientific
Vaara

221111000 221115000 221110051
ACRO221111000EA 28.5 EUR
ACRO221111000 ACRO221115000 ACRO221110051
Lyijy(II,IV)oksidi 98%
Lyijy(II,IV)oksidi
Kaava: Pb₃O₄
Kiehumispiste: 2682 °C (1013 hPa)
Tiheys: 9,1 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00078491
CAS-numero: 1314-41-6
EINECS: 215-235-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05449

Order Now

Testitulosten erittely

Appearance Orange Powder
Assay ≥97.5 % (PB3O4)
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Manganese (Mn) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR