4-Methoxyphenyl isocyanate 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 1-Isocyanato-4-methoxybenzene

218760050 218760250
ACRO218760050EA 71.1 EUR
ACRO218760050 ACRO218760250
4-Methoxyphenyl isocyanate 99%
4-Methoxyphenyl isocyanate
Kaava: C₈H₇NO₂
Kiehumispiste: 106…110 °C (16 mmHg)
Tiheys: 1,15 g/cm³
Leimahduspiste: 98 °C
MDL numero: MFCD00002026
CAS-numero: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR