Dikloorimetaani-D2 99.6+ atom % D for NMR spectroscopy

Toimittaja: Thermo Fisher Scientific
Varoitus

217350100
ACRO217350100EA 219 EUR
ACRO217350100
Dikloorimetaani-D2 99.6+ atom % D for NMR spectroscopy
Dikloorimetaani-D2

Packaged in 1.00 ml ampoules

Kaava: CD₂Cl₂
Kiehumispiste: 40 °C (1013 hPa)
Sulamispiste: –97 °C
Tiheys: 1,36 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000882
CAS-numero: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Water ≤0.04 % (Coulometric)
D-Enrichment ≥99.60 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR