Dikaliumvetyortofosfaatti, vedetön 99+% for analysis

Toimittaja: Thermo Fisher Scientific

215470010 215472500 215470050
ACRO215470010EA 144 EUR
ACRO215470010 ACRO215472500 ACRO215470050
Dikaliumvetyortofosfaatti, vedetön 99+% for analysis
Dikaliumvetyortofosfaatti
Kaava: K₂HPO₄
Sulamispiste: 340 °C
Tiheys: 2,3 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011383
CAS-numero: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Loss on drying ≤1 % (130°C)
pH 8.5 to 9.6 (5 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR