Metyyli-3-aminotiofeeni-2-karboksylaatti 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

214300100 214300500
ACRO214300100EA 60.2 EUR
ACRO214300100 ACRO214300500
Metyyli-3-aminotiofeeni-2-karboksylaatti 99%
Metyyli-3-aminotiofeeni-2-karboksylaatti
Kaava: C₆H₇NO₂S
Kiehumispiste: 100…102 °C (,1 mmHg)
Sulamispiste: 66…67 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009765
CAS-numero: 22288-78-4
EINECS: 244-895-8

Order Now

Testitulosten erittely

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 66 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR