Koboltti(II)sulfaatti-7 H2O 99+%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

213101000 213105000 213100025
ACRO213101000EA 91.3 EUR
ACRO213101000 ACRO213105000 ACRO213100025
Koboltti(II)sulfaatti-7 H2O 99+%, extra pure
Koboltti(II)sulfaatti-7 H2O
Kaava: CoSO₄·7H₂O
Sulamispiste: 98 °C
Tiheys: 2,03 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149658
CAS-numero: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 12,02510

Order Now

Testitulosten erittely

Appearance Red-brown Adhering crystalline powder or crystals
Titration Complexometric ≥99.0 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR