3-Anisoyylikloridi 99%

Toimittaja: Thermo Fisher Scientific
Vaara

212970050 212970250 212971000
ACRO212970050EA 38.1 EUR
ACRO212970050 ACRO212970250 ACRO212971000
3-Anisoyylikloridi 99%
3-Anisoyylikloridi
Kaava: C₈H₇ClO₂
Kiehumispiste: 123…125 °C (15 mmHg)
Tiheys: 1,214 g/cm³
Leimahduspiste: 92 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000673
CAS-numero: 1711-05-3
EINECS: 216-975-2
UN: 1729
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR