Alumiininitraatti-9 H2O 99.999% (trace metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

212240100 212241000
ACRO212240100EA 82.1 EUR
ACRO212240100 ACRO212241000
Alumiininitraatti-9 H2O 99.999% (trace metals basis)
Alumiininitraatti-9 H2O
Kaava: Al(NO₃)₃·9H₂O
Sulamispiste: 73 °C
Tiheys: 1,72 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149132
CAS-numero: 7784-27-2
ADR: 5.1,III

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 6.5 to 7.9 % (Al)
Total trace metal impurities ≤10 ppm

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR