Cetyldimethylethylammonium bromide 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

211871000 211875000
ACRO211871000EA 83.4 EUR
ACRO211871000 ACRO211875000
Cetyldimethylethylammonium bromide 99%
Cetyldimethylethylammonium bromide
Kaava: C₂₀H₄₄BrN
Sulamispiste: 190 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011774
CAS-numero: 124-03-8
EINECS: 204-672-8
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR