Bis(2,4-pentanedionato)molybdenum(VI) dioxide 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)

211140500 211142500
ACRO211140500EA 119 EUR
ACRO211140500 ACRO211142500
Bis(2,4-pentanedionato)molybdenum(VI) dioxide 97%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide

CAS No.: 17524-05-9

Kaava: C₁₀H₁₄MoO₆
Sulamispiste: 184 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011506
CAS-numero: 17524-05-9

Order Now

Testitulosten erittely

Appearance Yellow to grey-green to light brown Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR