Tetrapropyyliammoniumbromidi 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

209370250 209371000 209375000 209370025
ACRO209370250EA 23.4 EUR
ACRO209370250 ACRO209371000 ACRO209375000 ACRO209370025
Tetrapropyyliammoniumbromidi 98%
Tetrapropyyliammoniumbromidi
Kaava: C₁₂H₂₈BrN
Sulamispiste: 266…272 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011840
CAS-numero: 1941-30-6
EINECS: 217-727-6

Order Now

Testitulosten erittely

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR