Kupari(II)nitraatti-3 H2O 99% for analysis

Toimittaja: Thermo Fisher Scientific
Vaara

207681000 207685000 207680025
ACRO207681000EA 36.3 EUR
ACRO207681000 ACRO207685000 ACRO207680025
Kupari(II)nitraatti-3 H2O 99% for analysis
Kupari(II)nitraatti-3 H2O
Kaava: Cu(NO₃)₂·3H₂O
Sulamispiste: 114 °C
Tiheys: 2,05 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149671
CAS-numero: 10031-43-3
EINECS: 221-838-5
UN: 3085
ADR: 5.1,II
Merck Index: 13,02671

Order Now

Testitulosten erittely

Appearance Blue Powder or crystals and/or chunks
Titration Iodometric 98.5 to 104.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR