Retinoic acid 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

207341000 207340010 207340050
ACRO207341000EA 50.7 EUR
ACRO207341000 ACRO207340010 ACRO207340050
Retinoic acid 97%
Retinoic acid
Kaava: C₂₀H₂₈O₂
Sulamispiste: 179…184 °C
Storage Temperature: Pakastin
MDL numero: MFCD00001551
CAS-numero: 302-79-4
EINECS: 206-129-0

Order Now

Testitulosten erittely

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 184 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR