4-(Metyylitio)bentsaldehydi 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

207150500 207150050
ACRO207150500EA 164 EUR
ACRO207150500 ACRO207150050
4-(Metyylitio)bentsaldehydi 97%
4-(Metyylitio)bentsaldehydi
Kaava: C₈H₈OS
Kiehumispiste: 86…90 °C (1 mmHg)
Sulamispiste: 6 °C
Tiheys: 1,14 g/cm³
Leimahduspiste: >110 °C
MDL numero: MFCD00006948
CAS-numero: 3446-89-7
EINECS: 222-365-7
ADR: 9,

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6440 to 1.6500 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR