Divanadiumpentoksidi 99.6+%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Vanadiumpentoksidi

206422500 206420010
ACRO206422500EA 103 EUR
ACRO206422500 ACRO206420010
Divanadiumpentoksidi 99.6+%
Divanadiumpentoksidi
Kaava: O₅V₂
MW: 181.88 g/mol
Kiehumispiste: 1750 °C (760 mmHg)
Sulamispiste: 690 °C
Tiheys: 3.35 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011457
CAS-numero: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1 III
Merck Index: 13,09987

Order Now

Testitulosten erittely

Appearance Yellow to orange-brown Powder
Assay ≥99.60 %
Chromium (Cr) ≤100 ppm
Iron (Fe) ≤300 ppm
Potassium (K) ≤200 ppm
Silicon (Si) ≤200 ppm
Sodium (Na) ≤200 ppm

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR