Ammoniumheptamolybdaatti-4 H2O 99+% for analysis

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

205851000 205855000 205850025
ACRO205851000EA 53.3 EUR
ACRO205851000 ACRO205855000 ACRO205850025
Ammoniumheptamolybdaatti-4 H2O 99+% for analysis
Ammoniumheptamolybdaatti-4 H2O
Kaava: (NH₄)₆Mo₇O₂₄·4H₂O
Sulamispiste: 190 °C
Tiheys: 2,49 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00167059
CAS-numero: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Assay ≥99.0 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR