Rauta(II)kloriditetrahydraatii 99+%

Toimittaja: Thermo Fisher Scientific
Vaara

205080010 205080050 205082500 205080051
ACRO205080010EA 180 EUR
ACRO205080010 ACRO205080050 ACRO205082500 ACRO205080051
Rauta(II)kloriditetrahydraatii 99+%
Rauta(II)kloriditetrahydraatii
Kaava: FeCl₂·4H₂O
Sulamispiste: 105…110 °C
Tiheys: 1,926 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149709
CAS-numero: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Yellow to green Crystalline powder, crystals and/or chunks
Titration with KMnO4 ≥99.0 %
Sulfate (SO4) ≤200 ppm
Arsenic (As) ≤5 ppm
Copper (Cu) ≤50 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR