DL-Isocitric acid, trisodium salt hydrate 95%

Toimittaja: Thermo Fisher Scientific

205010010 205010050 205010250
ACRO205010010EA 79.2 EUR
ACRO205010010 ACRO205010050 ACRO205010250
DL-Isocitric acid, trisodium salt hydrate 95%
DL-Isocitric acid, trisodium salt hydrate

Appearance: White to off-white Crystalline powder

Kaava: C₆H₅Na₃O₇·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149284
CAS-numero: 1637-73-6

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤20 % (K.F.)
Titration with HClO4 ≥94 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR