4-Amino-N,N-dimetyylianiliini 2 HCl 99%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

204350250 204351000
ACRO204350250EA 124 EUR
ACRO204350250 ACRO204351000
4-Amino-N,N-dimetyylianiliini 2 HCl 99%
4-Amino-N,N-dimetyylianiliini 2 HCl
Kaava: (CH₃)₂NC₆H₄NH₂·2HCl
Sulamispiste: 222 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012991
CAS-numero: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280

Order Now

Testitulosten erittely

Appearance White to yellow to beige to pink to grey Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless to yellow to purple-brown
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR