(-)-Myrtenal 98%

Toimittaja: Thermo Fisher Scientific

203620100 203620500
ACRO203620100EA 70.3 EUR
ACRO203620100 ACRO203620500
(-)-Myrtenal 98%
(-)-Myrtenal

Appearance: Clear colorless to yellow Liquid

Kaava: C₁₀H₁₄O
Kiehumispiste: 220…221 °C (760 mmHg)
Tiheys: 0,98 g/cm³
Leimahduspiste: 78 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00074768
CAS-numero: 18486-69-6

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5010 to 1.5050 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -13° to -17° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR