Mangaanidioksidi 88%, electrolytically precipitated, active

Toimittaja: Thermo Fisher Scientific
Vaara

203191000 203195000 203190025
ACRO203191000EA 35.8 EUR
ACRO203191000 ACRO203195000 ACRO203190025
Mangaanidioksidi 88%, electrolytically precipitated, active
Mangaanidioksidi
Kaava: MnO₂
Sulamispiste: 535 °C (haj. lämpötila)
Tiheys: 5,03 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003463
CAS-numero: 1313-13-9
EINECS: 215-202-6
UN: 1479
ADR: 5.1,II
Merck Index: 14,05730

Order Now

Testitulosten erittely

Appearance Black Powder
Titration Iodometric ≥83 % (on dried substance)
Additional info Oxidation activity: >60% (yield)

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR