Bariumhydroksidioctahydraatii 98+% for analysis

Toimittaja: Thermo Fisher Scientific
Vaara

203140010 203142500 203140050
ACRO203140010EA 150 EUR
ACRO203140010 ACRO203142500 ACRO203140050
Bariumhydroksidioctahydraatii 98+% for analysis
Bariumhydroksidioctahydraatii
Kaava: Ba(OH)₂·8H₂O
Kiehumispiste: 780 °C (1013 hPa)
Sulamispiste: 78 °C
Tiheys: 2,18 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149152
CAS-numero: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Merck Index: 13,00980

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals or flakes
Titration with HCl ≥98.0 %
Chloride (Cl) ≤20 ppm
Barium carbonate ≤2 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Strontium (Sr) ≤1.5 %
Sulfur (S) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR