Vinyltrimethylsilane 97%

Toimittaja: Thermo Fisher Scientific
Vaara

200330250 200331000
ACRO200330250EA 79.3 EUR
ACRO200330250 ACRO200331000
Vinyltrimethylsilane 97%
Vinyltrimethylsilane
Kaava: C₅H₁₂Si
Kiehumispiste: 55 °C (1013 hPa)
Tiheys: 0,68 g/cm³
Leimahduspiste: –24 °C
MDL numero: MFCD00008606
CAS-numero: 754-05-2
EINECS: 212-042-9
UN: 1993

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3895 to 1.3915 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR