Litiumbromidi, vedetön 99+% for analysis

Toimittaja: Thermo Fisher Scientific
Varoitus

199871000 199875000 199870025 199870100
ACRO199871000EA 62.9 EUR
ACRO199871000 ACRO199875000 ACRO199870025 ACRO199870100
Litiumbromidi, vedetön 99+% for analysis
Litiumbromidi
Kaava: LiBr
Kiehumispiste: 1265 °C (760 mmHg)
Sulamispiste: 550 °C
Tiheys: 3,46 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011077
CAS-numero: 7550-35-8
EINECS: 231-439-8
Merck Index: 13,05548

Order Now

Testitulosten erittely

Appearance White Powder
Titration Argentometric ≥99.0 %
Loss on drying ≤0.5 % (400°C)
pH 6 to 12 (4 % at 20°C)
Chloride (Cl) ≤1500 ppm
Carbonate (CO3) ≤300 ppm
Sulfate (SO4) ≤200 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤3 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Potassium (K) ≤300 ppm
Sodium (Na) ≤400 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR