Cerium(IV)sulfaattitetrahydraatii 98+%

Toimittaja: Thermo Fisher Scientific
Vaara

199131000 199135000 199130025
ACRO199131000EA 199 EUR
ACRO199131000 ACRO199135000 ACRO199130025
Cerium(IV)sulfaattitetrahydraatii 98+%
Cerium(IV)sulfaattitetrahydraatii
Kaava: Ce(SO₄)₂·4H₂O
Sulamispiste: 180 - 200 °C
Tiheys: 3,91 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149427
CAS-numero: 10294-42-5
EINECS: 237-029-5
UN: 3260
ADR: 8,III
Merck Index: 12,02036

Order Now

Testitulosten erittely

Appearance Bright yellow Powder
Titration Iodometric ≥98.0 %
Chloride (Cl) ≤100 ppm
Copper (Cu) ≤30 ppm
Iron (Fe) ≤150 ppm
Lead (Pb) ≤30 ppm
Nickel (Ni) ≤30 ppm
Heavy metals (as Pb) ≤30 ppm
Insoluble matter ≤0.1 % (in H2SO4)
Subst not pptd by NH4OH ≤0.3 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR