Cerium(IV)oksidi 99.9% (trace metals basis)

Toimittaja: Thermo Fisher Scientific

199121000 199125000 199120025
ACRO199121000EA 63.2 EUR
ACRO199121000 ACRO199125000 ACRO199120025
Cerium(IV)oksidi 99.9% (trace metals basis)
Cerium(IV)oksidi
Kaava: CeO₂
Kiehumispiste: 3468 °C (1013 hPa)
Sulamispiste: 2600 °C
Tiheys: 7,13 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010933
CAS-numero: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now

Testitulosten erittely

Appearance White to yellow Powder (May give a pink to red-brown color due to traces lanthanum, neodymium, praseodymium, etc.)
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR