4-Fluoribentsamidi 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

197900050 197900250
ACRO197900050EA 39.8 EUR
ACRO197900050 ACRO197900250
4-Fluoribentsamidi 98%
4-Fluoribentsamidi
Kaava: C₇H₆FNO
Kiehumispiste: 287 °C (1013 hPa)
Sulamispiste: 154…158 °C
Tiheys: 1,43 g/cm³ (20 °C)
MDL numero: MFCD00007992
CAS-numero: 824-75-9
EINECS: 212-535-9

Order Now

Testitulosten erittely

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR