Bromi 99.6% for analysis

Toimittaja: Thermo Fisher Scientific
Vaara

196660010 196660250 196662500
ACRO196660010EA 249 EUR
ACRO196660010 ACRO196660250 ACRO196662500
Bromi 99.6% for analysis
Bromi
Kaava: Br₂
Kiehumispiste: 58,7 °C (760 mmHg)
Sulamispiste: –7,2 °C
Tiheys: 3,111 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010896
CAS-numero: 7726-95-6
EINECS: 231-778-1
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Testitulosten erittely

Appearance Dark red-brown Liquid
Titration Iodometric ≥99.5 %
Residue after evaporation ≤100 ppm
Chlorine (Cl) ≤500 ppm
Iodine (I) ≤10 ppm
Heavy metals (as Pb) ≤2 ppm
Sulfur compounds (as SO4) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR