Platina(IV)oksidi 83% Pt

Toimittaja: Thermo Fisher Scientific
Vaara

195320010 195320050
ACRO195320010EA 234 EUR
ACRO195320010 ACRO195320050
Platina(IV)oksidi 83% Pt
Platina(IV)oksidi
Kaava: O₂Pt
Sulamispiste: 450 °C
Tiheys: 11,8 g/cm³ (20 °C)
MDL numero: MFCD00011184
CAS-numero: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1,I
Merck Index: 12,07686

Order Now

Testitulosten erittely

Appearance Brown Powder
Assay ≥79 % (Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR